missing translation for 'onlineSavingsMsg'
Learn More

Trioctylmethylammonium Chloride (R=C8-C10) [Phase-transfer Catalyst], TCI America™

Catalog No. t082625g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T082625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T082625G Supplier TCI America Supplier No. T082625G
Only null left

Chemical Identifiers

CAS 63393-96-4
Molecular Formula C25H54ClN
Molecular Weight (g/mol) 404.16
MDL Number MFCD00011862
InChI Key XKBGEWXEAPTVCK-UHFFFAOYSA-M
Synonym methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride
PubChem CID 21218
ChEBI CHEBI:75286
IUPAC Name methyltrioctylazanium chloride
SMILES [Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC

Specifications

Color Yellow
Quantity 25 g
UN Number 2810
Physical Form Viscous Liquid
Chemical Name or Material Trioctylmethylammonium Chloride (R=C8-C10) [Phase-transfer Catalyst]
EINECSNumber (2)-0184&(9)-1971
RTECSNumber UZ2997500
TSCA Yes