missing translation for 'onlineSavingsMsg'
Learn More

Trimorpholinophosphine Oxide 98.0+%, TCI America™

Catalog No. p-7137391
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27485G 5 g
T274825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27485G Supplier TCI America Supplier No. T27485G
Only null left

Chemical Identifiers

CAS 4441-12-7
Molecular Formula C12H24N3O4P
Molecular Weight (g/mol) 305.32
MDL Number MFCD00047406
InChI Key WXMQHPKQCPCDQO-UHFFFAOYSA-N
Synonym trimorpholinophosphine oxide, 4,4',4-phosphoryltrimorpholine, tri 4-morpholinyl phosphine oxide, phosphoric acid trimorpholide, 4-bis morpholin-4-yl phosphoroso morpholine, tmpo;tris-morpholino-phosphine oxide, phosphoric trimorpholide, 5-n-propylbenzoic acid, acmc-2097fl, ksc493m7l
PubChem CID 78180
IUPAC Name 4-dimorpholin-4-ylphosphorylmorpholine
SMILES C1COCCN1P(=O)(N2CCOCC2)N3CCOCC3

Specifications

Melting Point 191°C
Color White
Quantity 5 g
Formula Weight 305.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Trimorpholinophosphine Oxide