missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyltetradecylammonium Chloride 98.0+%, TCI America™
Supplier: TCI America T0926500G
Specifications
| Trimethyltetradecylammonium Chloride | |
| White | |
| MFCD00059968 | |
| tetradecyltrimethylammonium chloride, myristyltrimethylammonium chloride, n,n,n-trimethyltetradecan-1-aminium chloride, trimethyltetradecylammonium chloride, 1-tetradecanaminium, n,n,n-trimethyl-, chloride, n-tetradecyltrimethylammonium chloride, rmv7nju3fx, unii-rmv7nju3fx, trimethyl tetradecyl azanium chloride, tetradecyl trimethyl ammonium chloride | |
| CCCCCCCCCCCCCC[N+](C)(C)C.[Cl-] | |
| 291.95 | |
| 291.95 | |
| Crystalline Powder |
| 4574-04-3 | |
| C17H38ClN | |
| 500 g | |
| CEYYIKYYFSTQRU-UHFFFAOYSA-M | |
| trimethyl(tetradecyl)azanium;chloride | |
| 20708 | |
| ≥98.0% (T) |
Chemical Identifiers
| 4574-04-3 | |
| 291.95 | |
| CEYYIKYYFSTQRU-UHFFFAOYSA-M | |
| 20708 | |
| CCCCCCCCCCCCCC[N+](C)(C)C.[Cl-] |
| C17H38ClN | |
| MFCD00059968 | |
| tetradecyltrimethylammonium chloride, myristyltrimethylammonium chloride, n,n,n-trimethyltetradecan-1-aminium chloride, trimethyltetradecylammonium chloride, 1-tetradecanaminium, n,n,n-trimethyl-, chloride, n-tetradecyltrimethylammonium chloride, rmv7nju3fx, unii-rmv7nju3fx, trimethyl tetradecyl azanium chloride, tetradecyl trimethyl ammonium chloride | |
| trimethyl(tetradecyl)azanium;chloride |
Safety and Handling
EINECSNumber : (2)-0184&(9)-1971
RTECSNumber : XB7985000
TSCA : Yes