missing translation for 'onlineSavingsMsg'
Learn More

Trimethyltetradecylammonium Chloride 98.0+%, TCI America™

Catalog No. T092625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T092625G 25 g
T0926500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T092625G Supplier TCI America Supplier No. T092625G

Chemical Identifiers

CAS 4574-04-3
Molecular Formula C17H38ClN
Molecular Weight (g/mol) 291.95
MDL Number MFCD00059968
InChI Key CEYYIKYYFSTQRU-UHFFFAOYSA-M
Synonym tetradecyltrimethylammonium chloride, myristyltrimethylammonium chloride, n,n,n-trimethyltetradecan-1-aminium chloride, trimethyltetradecylammonium chloride, 1-tetradecanaminium, n,n,n-trimethyl-, chloride, n-tetradecyltrimethylammonium chloride, rmv7nju3fx, unii-rmv7nju3fx, trimethyl tetradecyl azanium chloride, tetradecyl trimethyl ammonium chloride
PubChem CID 20708
IUPAC Name trimethyl(tetradecyl)azanium;chloride
SMILES CCCCCCCCCCCCCC[N+](C)(C)C.[Cl-]

Specifications

Color White
Quantity 25 g
Formula Weight 291.95
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethyltetradecylammonium Chloride
EINECSNumber (2)-0184&(9)-1971
RTECSNumber XB7985000
TSCA Yes