missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsulfonium tetrafluoroborate, 98+%
Description
Trimethylsulfonium tetrafluoroborate, is used as Methylene transfer agent and as a pharmaceutical intermediate. It is also used as a raw material in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trimethylsulfonium tetrafluoroborate, is used as Methylene transfer agent and as a pharmaceutical intermediate. It is also used as a raw material in organic synthesis.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Trimethylsulfonium tetrafluoroborate, is used as Methylene transfer agent and as a pharmaceutical intermediate. It is also used as a raw material in organic synthesis.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Specifications
| Melting Point | 214°C to 218°C |
| Quantity | 5 g |
| UN Number | UN3261 |
| Beilstein | 3708437 |
| Solubility Information | It is soluble in water. |
| Formula Weight | 163.98 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Trimethylsulfonium tetrafluoroborate |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 211-632-3 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only