missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfonium tetrafluoroborate, 98+%

Catalog No. AAL0064714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0064714 25 g
AAL0064706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0064714 Supplier Thermo Scientific Chemicals Supplier No. L0064714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 676-88-0 | C3H9BF4S | 163.97 g/mol

Trimethylsulfonium tetrafluoroborate, is used as Methylene transfer agent and as a pharmaceutical intermediate. It is also used as a raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsulfonium tetrafluoroborate, is used as Methylene transfer agent and as a pharmaceutical intermediate. It is also used as a raw material in organic synthesis.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 676-88-0
Molecular Formula C3H9BF4S
Molecular Weight (g/mol) 163.97
MDL Number MFCD00031582
InChI Key RNBNWTAKWJKQNN-UHFFFAOYSA-N
Synonym trimethylsulfonium tetrafluoroborate, trimethylsulphonium tetrafluoroborate 1-, trimethyl sulfonium fluoroborate, trimethylsulfanium tetrafluoroborate, trimethylsulphoniumtetrafluoroborate
PubChem CID 3083643
IUPAC Name trimethylsulfanium;tetrafluoroborate
SMILES C[S+](C)C.F[B-](F)(F)F

Specifications

Melting Point 214°C to 218°C
Quantity 25 g
UN Number UN3261
Beilstein 3708437
Solubility Information It is soluble in water.
Formula Weight 163.98
Percent Purity ≥98%
Chemical Name or Material Trimethylsulfonium tetrafluoroborate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 211-632-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only