missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsulfonium iodide, 98%

Catalog No. AAA1263930
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1263930 250 g
AAA1263918 50 g
AAA126390B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1263930 Supplier Thermo Scientific Chemicals Supplier No. A1263930

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2181-42-2 | C3H9IS | 204.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2181-42-2
Molecular Formula C3H9IS
Molecular Weight (g/mol) 204.069
MDL Number MFCD00011632
InChI Key VFJYIHQDILEQNR-UHFFFAOYSA-M
Synonym trimethylsulfonium iodide, trimethylsulphonium iodide, sulfonium, trimethyl-, iodide, trimethylsulfoniumiodide, trimethylsulfanium iodide, trimethyl-sulfonium iodide, unii-4z1liz2lun, 4z1liz2lun, sulfonium, trimethyl-, iodide 1:1, chembl25340
PubChem CID 75127
IUPAC Name trimethylsulfanium;iodide
SMILES C[S+](C)C.[I-]

Specifications

Melting Point ∼215°C (sublimation)
Quantity 250 g
Beilstein 3555192
Sensitivity Light sensitive
Formula Weight 204.07
Percent Purity 99%
Chemical Name or Material Trimethylsulfonium iodide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 218-555-4
RTECSNumber WR8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only