missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilylacetylene, 98%

Catalog No. p-3737685
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC203570050 5 g Glass bottle
AC203570250 25 g Glass bottle
AC203571000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC203570050 Supplier Thermo Scientific Chemicals Supplier No. 203570050
Only null left

CAS: 1066-54-2 | C5H10Si | 98.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1066-54-2
Molecular Formula C5H10Si
Molecular Weight (g/mol) 98.22
MDL Number MFCD00008569
InChI Key CWMFRHBXRUITQE-UHFFFAOYSA-N
Synonym trimethylsilylacetylene, trimethylsilyl acetylene, silane, ethynyltrimethyl, ethynyl-trimethyl-silane, ethynyl trimethyl silane, tms acetylene, ethynyltrimethyl silane, trimethylsilyl-acetylene, tmsacetylene
PubChem CID 66111
IUPAC Name ethynyl(trimethyl)silane
SMILES C[Si](C)(C)C#C

Specifications

Color Colorless
Density 0.6900g/mL
Boiling Point 53°C
Flash Point -34°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.3873 to 1.3903
Linear Formula (CH3)3SiC∽CH
Quantity 5 g
Specific Gravity 0.69
Solubility Information Solubility in water: reacts. Other solubilities: soluble in most common organic solvents
Formula Weight 98.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Trimethylsilylacetylene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-919-9

RUO – Research Use Only