missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7136472
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08715G 5 g
T087125G 25 g
T0871250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T08715G Supplier TCI America Supplier No. T08715G

Chemical Identifiers

CAS 27607-77-8
Molecular Formula C4H9F3O3SSi
Molecular Weight (g/mol) 222.253
MDL Number MFCD00000406
InChI Key FTVLMFQEYACZNP-UHFFFAOYSA-N
Synonym trimethylsilyl triflate, tmsotf, trifluoromethanesulfonic acid trimethylsilyl ester, trimethylsilyl trifluoromethylsulphonate, unii-z84v0cbh9j, methanesulfonic acid, trifluoro-, trimethylsilyl ester, trimethylsilyl trifluoromethylsulfonate, trimethylsllytrifluoromethanesulphonate, trimethylsilyl trifluoromethanesulphonate, trimethylsilyltrifluoromethanesulphonate
PubChem CID 65367
IUPAC Name trimethylsilyl trifluoromethanesulfonate
SMILES C[Si](C)(C)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
UN Number 2920
Formula Weight 222.25
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Trifluoromethanesulfonate [Trimethylsilylating Agent]
EINECSNumber (2)-3617
TSCA Yes