missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Trifluoroacetate 95.0+%, TCI America™

Catalog No. T26295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26295G 5 g
T262925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26295G Supplier TCI America Supplier No. T26295G

Chemical Identifiers

CAS 400-53-3
Molecular Formula C5H9F3O2Si
Molecular Weight (g/mol) 186.21
MDL Number MFCD00000413
InChI Key VIYXXANHGYSBLY-UHFFFAOYSA-N
Synonym trimethylsilyl trifluoroacetate, trimethylsilyltrifluoroacetate, acetic acid, trifluoro-, trimethylsilyl ester, acetic acid, 2,2,2-trifluoro-, trimethylsilyl ester, trifluoroacetic acid trimethylsilyl ester, tms trifluoroacetate, acmc-1ago5, cf3c o osi ch3 3, trimethylsilyl-trifluoroacetate, trifluoroacetic acid, tms derivative
PubChem CID 67863
IUPAC Name trimethylsilyl 2,2,2-trifluoroacetate
SMILES C[Si](C)(C)OC(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
UN Number 2924
Formula Weight 186.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Trifluoroacetate