missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl trifluoroacetate, 96%, Thermo Scientific™

Chemical Identifiers

CAS 400-53-3
Molecular Formula C5H9F3O2Si
Molecular Weight (g/mol) 186.21
MDL Number MFCD00000413
InChI Key VIYXXANHGYSBLY-UHFFFAOYSA-N
Synonym trimethylsilyl trifluoroacetate, trimethylsilyltrifluoroacetate, acetic acid, trifluoro-, trimethylsilyl ester, acetic acid, 2,2,2-trifluoro-, trimethylsilyl ester, trifluoroacetic acid trimethylsilyl ester, tms trifluoroacetate, acmc-1ago5, cf3c o osi ch3 3, trimethylsilyl-trifluoroacetate, trifluoroacetic acid, tms derivative
PubChem CID 67863
IUPAC Name trimethylsilyl 2,2,2-trifluoroacetate
SMILES C[Si](C)(C)OC(=O)C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC215970250
View Documents Promotion Details
N/A 25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

400-53-3
186.21
VIYXXANHGYSBLY-UHFFFAOYSA-N
67863
C[Si](C)(C)OC(=O)C(F)(F)F
C5H9F3O2Si
MFCD00000413
trimethylsilyl trifluoroacetate, trimethylsilyltrifluoroacetate, acetic acid, trifluoro-, trimethylsilyl ester, acetic acid, 2,2,2-trifluoro-, trimethylsilyl ester, trifluoroacetic acid trimethylsilyl ester, tms trifluoroacetate, acmc-1ago5, cf3c o osi ch3 3, trimethylsilyl-trifluoroacetate, trifluoroacetic acid, tms derivative
trimethylsilyl 2,2,2-trifluoroacetate

Specifications

400-53-3
88.0°C to 90.0°C
Authentic
C5H9F3O2Si
CF3CO2Si(CH3)3
04,IV,4004
trimethylsilyl trifluoroacetate, trimethylsilyltrifluoroacetate, acetic acid, trifluoro-, trimethylsilyl ester, acetic acid, 2,2,2-trifluoro-, trimethylsilyl ester, trifluoroacetic acid trimethylsilyl ester, tms trifluoroacetate, acmc-1ago5, cf3c o osi ch3 3, trimethylsilyl-trifluoroacetate, trifluoroacetic acid, tms derivative
VIYXXANHGYSBLY-UHFFFAOYSA-N
trimethylsilyl 2,2,2-trifluoroacetate
67863
96%
1.0800g/mL
0°C
95% min. (GC)
1.3350 to 1.3380
MFCD00000413
1.08
Solubility in water: reacts
C[Si](C)(C)OC(=O)C(F)(F)F
186.21
186.21
Trimethylsilyl trifluoroacetate, 96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away fr

GHS Signal Word: Danger

EINECSNumber : 206-923-7

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only