missing translation for 'onlineSavingsMsg'
Learn More

(Trimethylsilyl)methyllithium, 0.7M (10 wt%) solution in hexanes, AcroSeal™

CAS: 1822-00-0 | C4H11LiSi | 94.16 g/mol

$340.32 - $340.32

Specifications

Chemical Name or Material (Trimethylsilyl)methyllithium
Name Note 0.8M (12 wt%) solution in hexanes
CAS 1822-00-0,92112-69-1,92112-69-1
CAS Min % 7.0,86.0
CAS Max % 14.0,93.0
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC377451000
View Documents
Thermo Scientific Chemicals
377451000
100 mL
Each for $340.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Specifications

(Trimethylsilyl)methyllithium
1822-00-0,92112-69-1,92112-69-1
14.0,93.0
0.6500g/mL
−40°C
LiCH2Si(CH3)3
0.65
KVWLUDFGXDFFON-UHFFFAOYSA-N
lithium;methanidyl(trimethyl)silane
3482579
7 to 14 wt%
0.8M (12 wt%) solution in hexanes
7.0,86.0
Brown-Yellow to Colorless
65.0°C to 70.0°C
C4H11LiSi
MFCD00010747
trimethylsilyl methyllithium, trimethylsilyl methyl lithium, lich2sime3, pubchem18144, tms-ch2li, lithiomethyltrimethylsilane, acmc-1bvf0, trimethylsilylmethyl lithium
[Li+].C[Si](C)(C)[CH2-]
94.16
94.16
Solution
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only