missing translation for 'onlineSavingsMsg'
Learn More

(Trimethylsilyl)methyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. T15791G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15791G 1 g
T15795G 5 g
T157925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T15791G Supplier TCI America Supplier No. T15791G

Chemical Identifiers

CAS 64035-64-9
Molecular Formula C5H11F3O3SSi
Molecular Weight (g/mol) 236.28
MDL Number MFCD00009914
InChI Key VMDMAAJZSXXCQV-UHFFFAOYSA-N
Synonym trimethylsilyl methyl trifluoromethanesulfonate, trimethylsilylmethyl triflate, trimethylsilyl methyl triflate, trimethylsilylmethyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, trimethylsilyl methyl ester, acmc-1b9lz, trimethylsiylmethyltrifluoromethanesulfnate, trimethylsilylmethyltrifluoromethanesulfonate, trifluoromethanesulfonic acid trimethylsilylmethyl ester
PubChem CID 2760857
IUPAC Name trimethylsilylmethyl trifluoromethanesulfonate
SMILES C[Si](C)(C)COS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 168°C
Quantity 1 g
UN Number 2920
Formula Weight 236.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Trimethylsilyl)methyl Trifluoromethanesulfonate [Trimethylsilylmethylating Reagent]