missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsilyl Methacrylate (stabilized with N-Nitrosodiisopropanolamine) 98.0+%, TCI America™
Supplier: TCI America T35805ML
Specifications
| Trimethylsilyl Methacrylate (stabilized with N-Nitrosodiisopropanolamine) | |
| Colorless | |
| C7H14O2Si | |
| 5 mL | |
| trimethylsilyl methacrylate, 2-propenoic acid, 2-methyl-, trimethylsilyl ester, methacryloxytrimethylsilane, trimethylsilyl 2-methylpropenoate, methacrylic acid trimethylsilyl ester, acmc-20ajd2, trimethylsilylmethacrylate, methacrylic acid, tms derivative | |
| CC(=C)C(=O)O[Si](C)(C)C | |
| 158.272 | |
| 158.27 | |
| Liquid |
| 13688-56-7 | |
| 52°C | |
| MFCD00053868 | |
| 1993 | |
| PGQNYIRJCLTTOJ-UHFFFAOYSA-N | |
| trimethylsilyl 2-methylprop-2-enoate | |
| 2733839 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 13688-56-7 | |
| 158.272 | |
| PGQNYIRJCLTTOJ-UHFFFAOYSA-N | |
| 2733839 | |
| CC(=C)C(=O)O[Si](C)(C)C |
| C7H14O2Si | |
| MFCD00053868 | |
| trimethylsilyl methacrylate, 2-propenoic acid, 2-methyl-, trimethylsilyl ester, methacryloxytrimethylsilane, trimethylsilyl 2-methylpropenoate, methacrylic acid trimethylsilyl ester, acmc-20ajd2, trimethylsilylmethacrylate, methacrylic acid, tms derivative | |
| trimethylsilyl 2-methylprop-2-enoate |
Safety and Handling
EINECSNumber : (9)-1309
TSCA : Yes