missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Methacrylate (stabilized with BHT) 98.0+%, TCI America™

Catalog No. T32935ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32935ML 5 mL
T329325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32935ML Supplier TCI America Supplier No. T32935ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13688-56-7
Molecular Formula C7H14O2Si
Molecular Weight (g/mol) 158.272
MDL Number MFCD00053868
InChI Key PGQNYIRJCLTTOJ-UHFFFAOYSA-N
Synonym trimethylsilyl methacrylate, 2-propenoic acid, 2-methyl-, trimethylsilyl ester, methacryloxytrimethylsilane, trimethylsilyl 2-methylpropenoate, methacrylic acid trimethylsilyl ester, acmc-20ajd2, trimethylsilylmethacrylate, methacrylic acid, tms derivative
PubChem CID 2733839
IUPAC Name trimethylsilyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 52°C
Quantity 5 mL
UN Number 1993
Formula Weight 158.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Methacrylate (stabilized with BHT)
EINECSNumber (9)-1939
TSCA Yes