missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl isocyanate, 94%

Catalog No. AC224730050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC224730050 5 g Glass bottle
AC224730250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC224730050 Supplier Thermo Scientific Chemicals Supplier No. 224730050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1118-02-1 | C4H9NOSi | 115.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1118-02-1
Molecular Formula C4H9NOSi
Molecular Weight (g/mol) 115.21
MDL Number MFCD00001993
InChI Key NIZHERJWXFHGGU-UHFFFAOYSA-N
Synonym trimethylsilyl isocyanate, silane, isocyanatotrimethyl, trimethylsilylisocyanate, isocyanato trimethyl silane, trimethylisocyanatosilane, tmsisocyanate, tms isocyanate, tms-isocyanate, tms isocynate
PubChem CID 70696
SMILES C[Si](C)(C)N=C=O

Specifications

Melting Point -49.0°C
Color Colorless
Density 0.8500g/mL
Boiling Point 90.0°C to 92.0°C
Flash Point −2°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.3950 to 1.3970
Linear Formula (CH3)3SiNCO
Quantity 5 g
Beilstein 04, III, 1861
Fieser 06,634
Specific Gravity 0.85
IUPAC Name isocyanatotrimethylsilane
Formula Weight 115.21
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Trimethylsilyl isocyanate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
To
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable f
EINECSNumber 214-256-8
TSCA TSCA

RUO – Research Use Only