missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl isocyanate, 94%

Catalog No. AAA1263309
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1263309 10 g
AAA1263318 50 g
AAA1263330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1263309 Supplier Thermo Scientific Chemicals Supplier No. A1263309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1118-02-1 | C4H9NOSi | 115.21 g/mol

Trimethylsilyl isocyanate is used in the preparation of 1-unsubstituted 4-(dialkylamino) imidazolin-2-ones. It finds application in the carbamoylation of aromatic hydrocarbons and alcohols. It is also used in the preparation of primary amides from Grignard reagents. Further, it is used in the synthesis of cyclic imides. It is also utilized for the synthesis of trifluoroacetyl isocyanate and cyclic imides. In addition to this, it is employed in the conversion of isocyanates to carbodiimides using cyclopentadienyl Mn(CO)3 catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl isocyanate is used in the preparation of 1-unsubstituted 4-(dialkylamino) imidazolin-2-ones. It finds application in the carbamoylation of aromatic hydrocarbons and alcohols. It is also used in the preparation of primary amides from Grignard reagents. Further, it is used in the synthesis of cyclic imides. It is also utilized for the synthesis of trifluoroacetyl isocyanate and cyclic imides. In addition to this, it is employed in the conversion of isocyanates to carbodiimides using cyclopentadienyl Mn(CO)3 catalysis.

Solubility
Miscible with ethers and o-dichlorobenzene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids, strong bases, alcohols and amines.

Chemical Identifiers

CAS 1118-02-1
Molecular Formula C4H9NOSi
Molecular Weight (g/mol) 115.21
MDL Number MFCD00001993
InChI Key NIZHERJWXFHGGU-UHFFFAOYSA-N
Synonym trimethylsilyl isocyanate, silane, isocyanatotrimethyl, trimethylsilylisocyanate, isocyanato trimethyl silane, trimethylisocyanatosilane, tmsisocyanate, tms isocyanate, tms-isocyanate, tms isocynate
PubChem CID 70696
IUPAC Name isocyanato(trimethyl)silane
SMILES C[Si](C)(C)N=C=O

Specifications

Melting Point -49°C
Density 0.859
Boiling Point 90°C to 92°C
Flash Point −2°C (28°F)
Odor Acrid
Refractive Index 1.396
Quantity 10 g
UN Number UN2478
Beilstein 1744962
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Miscible with ethers and o-dichlorobenzene.
Formula Weight 115.21
Percent Purity 94%
Chemical Name or Material Trimethylsilyl isocyanate
Show More Show Less
Hazard Category H225-H315-H319-H331-H334-H335
Hazard Statement GHS H Statement
H225-H330-H334-H315-H319-H335
Highly flammable liquid and vapour.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P311-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOCYANATES, FLAMMABLE, TOXIC, N.O.S.
EINECSNumber 214-256-8
TSCA Yes
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only