missing translation for 'onlineSavingsMsg'
Learn More

(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes

CAS: 18107-18-1 | C4H10N2Si | 114.22 g/mol

Supplier:  Thermo Scientific Chemicals 385330250

 View more versions of this product

Catalog No. AC385330250

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Specifications

(Trimethylsilyl)diazomethane
32.15,64.47
Yellow
96.0°C
Glass bottle
(CH3)3SiCHN2
0.675
Solubility in water: insoluble
C[Si](C)(C)C=[N+]=[N-]
114.22
114.22
Solution
18107-18-1,92112-69-1
35.54,67.85
0.6750g/mL
−23°C
C4H10N2Si
25 mL
trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4
ONDSBJMLAHVLMI-UHFFFAOYSA-N
diazomethyl(trimethyl)silane
167693
Technical

Chemical Identifiers

18107-18-1
114.22
trimethylsilyl diazomethane, trimethylsilyldiazomethane, diazomethyl trimethylsilane, tms-diazomethane, diazomethyl trimethyl silane, tmschn2, silane, diazomethyl trimethyl, unii-qi98hqo8c4, diazomethyl-trimethyl-silane, qi98hqo8c4
diazomethyl(trimethyl)silane
C4H10N2Si
ONDSBJMLAHVLMI-UHFFFAOYSA-N
167693
C[Si](C)(C)C=[N+]=[N-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Causes skin irritation.
Suspected of damaging fertility.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Obtain special instructions before use.
Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot su

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only