Learn More
Trimethylsilyl cyanide, 98%, AcroSeal™
CAS: 7677-24-9 | C4H9NSi | 99.21 g/mol
Supplier: Thermo Scientific Chemicals 422061000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Trimethylsilyl cyanide | |
| 11.0°C | |
| 114.0°C to 117.0°C | |
| 97.5% min. (GC) | |
| C4H9NSi | |
| (CH3)3SiCN | |
| 100 mL | |
| 0.744 | |
| Solubility in water: reacts | |
| C[Si](C)(C)C#N | |
| 99.21 | |
| 99.21 |
| 7677-24-9 | |
| 0.7440g/mL | |
| 1°C | |
| AcroSeal™ Glass bottle | |
| 1.3880 to 1.3920 | |
| MFCD00001765 | |
| 04,542; 05,720; 06,632; 07,397; 08,133; 09,127; 10,112; 11,147; 12,148; 13,87; 14,107; 15,102; 17,89 | |
| trimethylsilyl cyanide, cyanotrimethylsilane, trimethylsilanecarbonitrile, trimethylsilylcyanide, silanecarbonitrile, trimethyl, trimethylsilylnitrile, tmscn, trimethyl silane cyanide, trimethylsilylcarbonitrile, tms cyanide | |
| LEIMLDGFXIOXMT-UHFFFAOYSA-N | |
| trimethylsilylformonitrile | |
| 82115 | |
| 98% |
Chemical Identifiers
| 7677-24-9 | |
| 99.21 | |
| LEIMLDGFXIOXMT-UHFFFAOYSA-N | |
| 82115 | |
| C[Si](C)(C)C#N |
| C4H9NSi | |
| MFCD00001765 | |
| trimethylsilyl cyanide, cyanotrimethylsilane, trimethylsilanecarbonitrile, trimethylsilylcyanide, silanecarbonitrile, trimethyl, trimethylsilylnitrile, tmscn, trimethyl silane cyanide, trimethylsilylcarbonitrile, tms cyanide | |
| trimethylsilylformonitrile |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal in contact with skin.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
GHS Signal Word: Danger
EINECSNumber : 231-657-3
RUO – Research Use Only