missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Trimethylsilyl cyanide

Catalog No. 21284925G
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21284925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21284925G Supplier Sigma Aldrich Supplier No. 21284925G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylsilyl cyanide

Specifications

CAS 7677-24-9
Melting Point 8°C to 11°C (lit.)
Density 0.793 g/mL (at 20°C (literature))
Boiling Point 114°C to 117°C (lit.)
Molecular Formula C4H9NSi
Refractive Index n20/D 1.392 (literature)
Linear Formula (CH3)3 SiCN
MDL Number MFCD00001765
Quantity 25 g
Synonym Cyanotrimethylsilane; TMSCN
Molecular Weight (g/mol) 99.21
Percent Purity 98%
Show More Show Less