missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Bromoacetate 97.0+%, TCI America™

Catalog No. B15785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15785G 5 g
B157825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15785G Supplier TCI America Supplier No. B15785G
Only null left

Chemical Identifiers

CAS 18291-80-0
Molecular Formula C5H11BrO2Si
Molecular Weight (g/mol) 211.13
MDL Number MFCD00000187
InChI Key ZCHHFMWUDHXPFN-UHFFFAOYSA-N
Synonym trimethylsilyl bromoacetate, bromoacetic acid trimethylsilyl ester, acetic acid, bromo-, trimethylsilyl ester, acmc-209ek0, trimethylsilyl monobromoacetate, trimethylsilylbromoacetate, bromoacetic acid, tms derivative, zchhfmwudhxpfn-uhfffaoysa
PubChem CID 87554
IUPAC Name trimethylsilyl 2-bromoacetate
SMILES C[Si](C)(C)OC(=O)CBr

Specifications

Color Yellow
Boiling Point 83°C
Quantity 5 g
UN Number 2920
Formula Weight 211.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Bromoacetate