missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Azide 95.0+%, TCI America™

Catalog No. p-7136437
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08015G 5 g
T080125G 25 g
T0801100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T08015G Supplier TCI America Supplier No. T08015G
Only null left

Chemical Identifiers

CAS 4648-54-8
Molecular Formula C3H9N3Si
Molecular Weight (g/mol) 115.21
MDL Number MFCD00001986
InChI Key SEDZOYHHAIAQIW-UHFFFAOYSA-N
Synonym trimethylsilyl azide, silane, azidotrimethyl, trimethylsilylazide, azido trimethyl silane, ccris 8048, tms azide, tms-azide, trimethyl silylazide, trimethylsilyl-azide, azido trimethylsilane
PubChem CID 78378
IUPAC Name azidotrimethylsilane
SMILES C[Si](C)(C)N=[N+]=[N-]

Specifications

Color Yellow
Boiling Point 97°C
Quantity 5 g
UN Number 1992
Formula Weight 115.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Azide
TSCA Yes
Recommended Storage Refrigerator