missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl azide, 94%

Catalog No. AAL0017306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0017306 5 g
AAL0017314 25 g
AAL0017322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0017306 Supplier Thermo Scientific Chemicals Supplier No. L0017306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4648-54-8 | C3H9N3Si | 115.21 g/mol

Trimethylsilyl azide is used as an azidonation reagent for amines, amides, aldehydes and ketones. It is involved in the preparation of alfa- and beta-siloxy azides from carbonyl compounds and epoxides. It is also used in the preparation of heterocyclic compounds and also acts as an effective substitute for hydrazoic acid. Further, it is used in the Oseltamivir total synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl azide is used as an azidonation reagent for amines, amides, aldehydes and ketones. It is involved in the preparation of alfa- and beta-siloxy azides from carbonyl compounds and epoxides. It is also used in the preparation of heterocyclic compounds and also acts as an effective substitute for hydrazoic acid. Further, it is used in the Oseltamivir total synthesis.

Solubility
Miscible with toluene, dichloromethane, diethyl ether and most organic solvents.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 4648-54-8
Molecular Formula C3H9N3Si
Molecular Weight (g/mol) 115.21
MDL Number MFCD00001986
InChI Key SEDZOYHHAIAQIW-UHFFFAOYSA-N
Synonym trimethylsilyl azide, silane, azidotrimethyl, trimethylsilylazide, azido trimethyl silane, ccris 8048, tms azide, tms-azide, trimethyl silylazide, trimethylsilyl-azide, azido trimethylsilane
PubChem CID 78378
SMILES C[Si](C)(C)N=[N+]=[N-]

Specifications

Melting Point -95°C
Density 0.872
Boiling Point 95°C to 99°C
Flash Point 6°C (42°F)
Refractive Index 1.414
Quantity 5 g
UN Number UN1992
Beilstein 1903730
Sensitivity Moisture sensitive
Solubility Information Miscible with toluene,dichloromethane,diethyl ether and most organic solvents.
IUPAC Name azidotrimethylsilane
Formula Weight 115.21
Percent Purity 94%
Chemical Name or Material Trimethylsilyl azide
Show More Show Less
Hazard Category H225-H301+H311+H331
Hazard Statement GHS H Statement
H225-H300-H310-H330
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 225-078-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only