missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Acetate 98.0+%, TCI America™

Catalog No. p-7117519
Change view
Click to view available options
Quantity:
25 mL
250 mL
Catalog No. Quantity
A156725ML 25 mL
A1567250ML 250 mL
2 options

Catalog No. A156725ML

Supplier: TCI America A156725ML

Only null left

Chemical Identifiers

CAS 2754-27-0
Molecular Formula C5H12O2Si
Molecular Weight (g/mol) 132.234
MDL Number MFCD00008695
InChI Key QHUNJMXHQHHWQP-UHFFFAOYSA-N
Synonym acetoxytrimethylsilane, trimethylacetoxysilane, acetato trimethylsilane, acetic acid trimethylsilyl ester, silanol, trimethyl-, acetate, silanol, 1,1,1-trimethyl-, 1-acetate, tmsoac, o-trimethylsilylacetate, trimethylsilyl acetate
PubChem CID 75988
IUPAC Name trimethylsilyl acetate
SMILES CC(=O)O[Si](C)(C)C

Specifications

Melting Point -32°C
Color Colorless
Boiling Point 108°C
Quantity 25 mL
UN Number 1993
Formula Weight 132.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Acetate
EINECSNumber (9)-1939
TSCA Yes