missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl Acetate 98.0+%, TCI America™

Catalog No. A156725ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A156725ML 25 mL
A1567250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A156725ML Supplier TCI America Supplier No. A156725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2754-27-0
Molecular Formula C5H12O2Si
Molecular Weight (g/mol) 132.234
MDL Number MFCD00008695
InChI Key QHUNJMXHQHHWQP-UHFFFAOYSA-N
Synonym acetoxytrimethylsilane, trimethylacetoxysilane, acetato trimethylsilane, acetic acid trimethylsilyl ester, silanol, trimethyl-, acetate, silanol, 1,1,1-trimethyl-, 1-acetate, tmsoac, o-trimethylsilylacetate, trimethylsilyl acetate
PubChem CID 75988
IUPAC Name trimethylsilyl acetate
SMILES CC(=O)O[Si](C)(C)C

Specifications

Melting Point -32°C
Color Colorless
Boiling Point 108°C
Quantity 25 mL
UN Number 1993
Formula Weight 132.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylsilyl Acetate
EINECSNumber (9)-1939
TSCA Yes