missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilanol, 95%

CAS: 1066-40-6 | C3H10OSi| 90.20 g/mol

Supplier:  Thermo Scientific Chemicals H6127606

 View more versions of this product

Catalog No. AAH6127606


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Trimethylsilanol used as a hydrophobic coat on silicate surfaces. Used as an as an antimicrobial agent.

Solubility
Soluble in water (1.00 mg/ml).

Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications

Specifications

98°C to 100°C
1066-40-6
C3H9LiOSi
MFCD02751657
1361356
trimethylsilanol, silanol, trimethyl, unii-z4bin3300p, trimethylhydroxysilane, ccris 1320, hydroxy trimethyl silane, silanol, 1,1,1-trimethyl, ksc504m9b, 2004-14-0 lithium salt, 10519-96-7 potassium salt
OXOZHAWWRPCVGL-UHFFFAOYSA-N
hydroxy(trimethyl)silane
66110
95%
Trimethylsilanol
5 g
(CH3)3SiOH
UN1993
4°C (39°F)
Soluble in water (1.00mg/ml).
[Li+].C[Si](C)(C)[O-]
96.13
90.2
0.814

Chemical Identifiers

1066-40-6
96.13
OXOZHAWWRPCVGL-UHFFFAOYSA-N
66110
[Li+].C[Si](C)(C)[O-]
C3H9LiOSi
MFCD02751657
trimethylsilanol, silanol, trimethyl, unii-z4bin3300p, trimethylhydroxysilane, ccris 1320, hydroxy trimethyl silane, silanol, 1,1,1-trimethyl, ksc504m9b, 2004-14-0 lithium salt, 10519-96-7 potassium salt
hydroxy(trimethyl)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 213-914-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only