missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilanol, 95%

Catalog No. AAH6127622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6127622 100 g
AAH6127606 5 g
AAH6127614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6127622 Supplier Thermo Scientific Chemicals Supplier No. H6127622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1066-40-6 | C3H10OSi| 90.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Trimethylsilanol used as a hydrophobic coat on silicate surfaces. Used as an as an antimicrobial agent.

Solubility
Soluble in water (1.00 mg/ml).

Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 1066-40-6
Molecular Formula C3H9LiOSi
Molecular Weight (g/mol) 96.13
MDL Number MFCD02751657
InChI Key OXOZHAWWRPCVGL-UHFFFAOYSA-N
Synonym trimethylsilanol, silanol, trimethyl, unii-z4bin3300p, trimethylhydroxysilane, ccris 1320, hydroxy trimethyl silane, silanol, 1,1,1-trimethyl, ksc504m9b, 2004-14-0 lithium salt, 10519-96-7 potassium salt
PubChem CID 66110
IUPAC Name hydroxy(trimethyl)silane
SMILES [Li+].C[Si](C)(C)[O-]

Specifications

Boiling Point 98°C to 100°C
Quantity 100 g
Flash Point 4°C (39°F)
Density 0.814
Linear Formula (CH3)3SiOH
UN Number UN1993
Beilstein 1361356
Solubility Information Soluble in water (1.00mg/ml).
Formula Weight 90.2
Percent Purity 95%
Chemical Name or Material Trimethylsilanol
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 213-914-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only