missing translation for 'onlineSavingsMsg'
Learn More

Trimethylpyruvic acid, ca. 60% aq. soln.

Catalog No. p-7028329
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2558218 50 g
AAB2558230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2558218 Supplier Thermo Scientific Chemicals Supplier No. B2558218

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 815-17-8 | C6H10O3 | 130.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 815-17-8
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00154352
InChI Key IAWVHZJZHDSEOC-UHFFFAOYSA-N
Synonym 3,3-dimethyl-2-oxobutyric acid, trimethylpyruvic acid, butanoic acid, 3,3-dimethyl-2-oxo, pyruvic acid, trimethyl, glyoxylic acid, tert-butyl, unii-29cl615clh, 3,3-dimethyl-2-oxo-butanoic acid, butyric acid, 3,3-dimethyl-2-oxo, tert-butylglyoxylic acid, trimethlpyruvic acid
PubChem CID 13150
IUPAC Name 3,3-dimethyl-2-oxobutanoic acid
SMILES CC(C)(C)C(=O)C(=O)O

Specifications

Linear Formula (CH3)3CCOCO2H
Quantity 50 g
UN Number UN3265
Sensitivity May form a precipitate
Formula Weight 130.14
Concentration or Composition (by Analyte or Components) ≈60% aq. Soln.
Chemical Name or Material Trimethylpyruvic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-418-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only