missing translation for 'onlineSavingsMsg'
Learn More

Trimethylpropylammonium Bromide 98.0+%, TCI America™

Catalog No. T276225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T276225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T276225G Supplier TCI America Supplier No. T276225G
Only null left

Chemical Identifiers

CAS 2650-50-2
Molecular Formula C6H16BrN
Molecular Weight (g/mol) 182.105
MDL Number MFCD06227832
InChI Key FCGQIZKUTMUWDC-UHFFFAOYSA-M
PubChem CID 75853
IUPAC Name trimethyl(propyl)azanium;bromide
SMILES CCC[N+](C)(C)C.[Br-]

Specifications

Melting Point 238°C
Color White-Yellow
Quantity 25 g
Formula Weight 182.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylpropylammonium Bromide