missing translation for 'onlineSavingsMsg'
Learn More

Trimethylphenylammonium Tribromide 98.0+%, TCI America™

Catalog No. p-7134849
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P092825G 25 g
P0928500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P092825G Supplier TCI America Supplier No. P092825G
Only null left

Chemical Identifiers

CAS 4207-56-1
Molecular Formula C9H14I3N
Molecular Weight (g/mol) 516.931
MDL Number MFCD00011789
InChI Key RHXJPCQWYPXLEH-UHFFFAOYSA-N
Synonym mono n,n,n-trimethylbenzenaminium tribromide
PubChem CID 21525000
IUPAC Name trimethyl(phenyl)azanium;triiodide
SMILES C[N+](C)(C)C1=CC=CC=C1.I[I-]I

Specifications

Melting Point 115°C
Quantity 25 g
UN Number 1759
Formula Weight 375.93
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylphenylammonium Tribromide
EINECSNumber (9)-1187
TSCA Yes