missing translation for 'onlineSavingsMsg'
Learn More

Trimethylphenylammonium Iodide 98.0+%, TCI America™

Catalog No. P024625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P024625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P024625G Supplier TCI America Supplier No. P024625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-04-4
Molecular Formula C9H14IN
Molecular Weight (g/mol) 263.122
MDL Number MFCD00011791
InChI Key KKLAORVGAKUOPZ-UHFFFAOYSA-M
Synonym phenyltrimethylammonium iodide, trimethylphenylammonium iodide, n,n,n-trimethylanilinium iodide, trimethylanilinium iodide, trimethylanillinium iodide, n,n-dimethylaniline methiodide, unii-4o888xq7fp, ammonium, phenyltrimethyl-, iodide, n,n,n-trimethylbenzenaminium iodide, benzenaminium, n,n,n-trimethyl-, iodide
PubChem CID 94135
IUPAC Name trimethyl(phenyl)azanium;iodide
SMILES C[N+](C)(C)C1=CC=CC=C1.[I-]

Specifications

Melting Point 227°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 263.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylphenylammonium Iodide
EINECSNumber (9)-1187
RTECSNumber BT2450000
TSCA Yes