missing translation for 'onlineSavingsMsg'
Learn More

Trimethylphenylammonium Chloride 98.0+%, TCI America™

Catalog No. P024425G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P024425G 25 g
P0244100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P024425G Supplier TCI America Supplier No. P024425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 138-24-9
Molecular Formula C9H14ClN
Molecular Weight (g/mol) 171.67
MDL Number MFCD00011790
InChI Key MQAYPFVXSPHGJM-UHFFFAOYSA-M
Synonym phenyltrimethylammonium chloride, n,n,n-trimethylbenzenaminium chloride, trimethylphenylammonium chloride, n,n,n-trimethylanilinium chloride, ammonyx 200, benzenaminium, n,n,n-trimethyl-, chloride, trimethylphenylammoniumchloride, trimethylanilinium chloride, ammonium, phenyltrimethyl-, chloride, trimethylphenyl ammonium chloride
PubChem CID 67309
IUPAC Name N,N,N-trimethylanilinium chloride
SMILES [Cl-].C[N+](C)(C)C1=CC=CC=C1

Specifications

Color White
Quantity 25 g
UN Number 2811
Formula Weight 171.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylphenylammonium Chloride
EINECSNumber (9)-1187
RTECSNumber BT2190000
TSCA Yes