missing translation for 'onlineSavingsMsg'
Learn More

Trimethylolpropane Trimethacrylate (stabilized with MEHQ) 90.0+%, TCI America™

$42.87 - $124.89

Chemical Identifiers

CAS 3290-92-4
Molecular Formula C18H26O6
Molecular Weight (g/mol) 338.4
MDL Number MFCD00008588
InChI Key OKKRPWIIYQTPQF-UHFFFAOYSA-N
Synonym 2-Ethyl-2-hydroxymethyl-1,3-propanediol Trimethacrylate
PubChem CID 18689
ChEBI CHEBI:35029
IUPAC Name 2,2-bis(2-methylprop-2-enoyloxymethyl)butyl 2-methylprop-2-enoate
SMILES CCC(COC(=O)C(=C)C)(COC(=O)C(=C)C)COC(=O)C(=C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T091225ML
View Documents
TCI America
T091225ML
25 mL N/A
Only null left
 
T0912500ML
View Documents
TCI America
T0912500ML
500 mL N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3290-92-4
338.4
OKKRPWIIYQTPQF-UHFFFAOYSA-N
18689
2,2-bis(2-methylprop-2-enoyloxymethyl)butyl 2-methylprop-2-enoate
C18H26O6
MFCD00008588
2-Ethyl-2-hydroxymethyl-1,3-propanediol Trimethacrylate
CHEBI:35029
CCC(COC(=O)C(=C)C)(COC(=O)C(=C)C)COC(=O)C(=C)C

Specifications

3290-92-4
Yellow
C18H26O6
25 mL
OKKRPWIIYQTPQF-UHFFFAOYSA-N
2,2-bis(2-methylprop-2-enoyloxymethyl)butyl 2-methylprop-2-enoate
18689
338.40
Liquid
-14°C
155°C
MFCD00008588
2-Ethyl-2-hydroxymethyl-1,3-propanediol Trimethacrylate
CCC(COC(=O)C(=C)C)(COC(=O)C(=C)C)COC(=O)C(=C)C
338.4
CHEBI:35029
≥90.0% (GC)
Trimethylolpropane Trimethacrylate (stabilized with MEHQ)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0769&(2)-1062

RTECSNumber : TY6675000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'