missing translation for 'onlineSavingsMsg'
Learn More

Trimethylolpropane Triacrylate (stabilized with MEHQ) 75.0+%, TCI America™

Catalog No. T0949500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0949500G 500 g
T094925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0949500G Supplier TCI America Supplier No. T0949500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15625-89-5
Molecular Formula C15H20O6
Molecular Weight (g/mol) 296.32
MDL Number MFCD00008628
InChI Key DAKWPKUUDNSNPN-UHFFFAOYSA-N
Synonym trimethylolpropane triacrylate, tmpta, viscoat 295, nk ester a tmpt, sartomer sr 351, ogumont t 200, saret 351, setalux uv 2241, ccris 92, unii-4b67kgl96s
PubChem CID 27423
ChEBI CHEBI:35028
IUPAC Name 2,2-bis[(prop-2-enoyloxy)methyl]butyl prop-2-enoate
SMILES CCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C

Specifications

Color Yellow
Quantity 500 g
Formula Weight 296.32
Percent Purity ≥75.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethylolpropane Triacrylate (stabilized with MEHQ)
EINECSNumber (2)-1010&(2)-0958
RTECSNumber AT4810000
TSCA Yes