missing translation for 'onlineSavingsMsg'
Learn More

Trimethylolpropane 98.0+%, TCI America™

Catalog No. T048025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T048025G 25 g
T0480500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T048025G Supplier TCI America Supplier No. T048025G

Chemical Identifiers

CAS 77-99-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00004694
InChI Key ZJCCRDAZUWHFQH-UHFFFAOYSA-N
Synonym trimethylolpropane, 2-ethyl-2-hydroxymethyl propane-1,3-diol, ethriol, 2-ethyl-2-hydroxymethyl-1,3-propanediol, hexaglycerine, ettriol, etriol, trimethylol propane, tmp alcohol, 1,1,1-tris hydroxymethyl propane
PubChem CID 6510
IUPAC Name 2-ethyl-2-(hydroxymethyl)propane-1,3-diol
SMILES CCC(CO)(CO)CO

Specifications

Melting Point 58°C
Color White
Boiling Point 149°C
Quantity 25 g
Formula Weight 134.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylolpropane
EINECSNumber (2)-0245
RTECSNumber TY6470000
TSCA Yes