missing translation for 'onlineSavingsMsg'
Learn More

Trimethylolpropane Phosphite 94.0+%, TCI America™

Catalog No. p-7136442
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08165G 5 g
T081625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08165G Supplier TCI America Supplier No. T08165G
Only null left

Chemical Identifiers

CAS 824-11-3
Molecular Formula C6H11O3P
Molecular Weight (g/mol) 162.125
MDL Number MFCD00152500
InChI Key QRUSNTDXJQBKBI-UHFFFAOYSA-N
Synonym 4-Ethyl-1-phospha-2,6,7-trioxabicyclo[2.2.2]octane
PubChem CID 13210
IUPAC Name 4-ethyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane
SMILES CCC12COP(OC1)OC2

Specifications

Melting Point 56°C
Color White
Boiling Point 100°C
Quantity 5 g
UN Number 3464
Formula Weight 162.12
Percent Purity ≥94.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylolpropane Phosphite
EINECSNumber (2)-1909
RTECSNumber TY6650000
TSCA Yes