missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylolpropane Phosphite 94.0+%, TCI America™
Specifications
Specifications
| Melting Point | 56°C |
| Color | White |
| Boiling Point | 100°C |
| Quantity | 5 g |
| UN Number | 3464 |
| Formula Weight | 162.12 |
| Percent Purity | ≥94.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Trimethylolpropane Phosphite |
Safety and Handling
| EINECSNumber | (2)-1909 |
| RTECSNumber | TY6650000 |
| TSCA | Yes |