missing translation for 'onlineSavingsMsg'
Learn More

Trimethylolethane 98.0+%, TCI America™

Catalog No. p-7136305
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T047925G 25 g
T0479500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T047925G Supplier TCI America Supplier No. T047925G
Only null left

Chemical Identifiers

CAS 77-85-0
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.148
MDL Number MFCD00004687
InChI Key QXJQHYBHAIHNGG-UHFFFAOYSA-N
Synonym trimethylolethane, 1,1,1-tris hydroxymethyl ethane, trimet, methriol, metriol, pentaglycerine, pentaglycerol, methyltrimethanolmethane, 2-hydroxymethyl-2-methylpropane-1,3-diol, tris hydroxymethyl ethane
PubChem CID 6502
IUPAC Name 2-(hydroxymethyl)-2-methylpropane-1,3-diol
SMILES CC(CO)(CO)CO

Specifications

Melting Point 201°C
Color White
Boiling Point 137°C
Quantity 25 g
Formula Weight 120.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylolethane
EINECSNumber (2)-0243
RTECSNumber TY7137500
TSCA Yes