missing translation for 'onlineSavingsMsg'
Learn More

Trimethylnonylammonium Bromide 98.0+%, TCI America™

Catalog No. T27345G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27345G Supplier TCI America Supplier No. T27345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1943-11-9
Molecular Formula C12H28BrN
Molecular Weight (g/mol) 266.27
MDL Number MFCD00041986
InChI Key SSEGNMJSEROZIF-UHFFFAOYSA-M
Synonym Nonyltrimethylammonium Bromide
PubChem CID 74750
IUPAC Name trimethyl(nonyl)azanium;bromide
SMILES CCCCCCCCC[N+](C)(C)C.[Br-]

Specifications

Melting Point 223°C
Color White
Quantity 5 g
Formula Weight 266.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylnonylammonium Bromide
EINECSNumber (2)-0184&(9)-1971
TSCA Yes