missing translation for 'onlineSavingsMsg'
Learn More

Trimethylhexamethylenediamine (2,2,4- and 2,4,4- mixture) 99.0+%, TCI America™

Catalog No. T060025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T060025ML 25 mL
T0600500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T060025ML Supplier TCI America Supplier No. T060025ML
Only null left

Chemical Identifiers

CAS 25513-64-8
Molecular Formula C9H22N2
MDL Number MFCD00059849

Specifications

Melting Point -80°C
Color Yellow
pH 11.6 (8g/L H2O soln. 20°C)
Boiling Point 232°C
Quantity 25 mL
UN Number 2327
Formula Weight 158.29
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Trimethylhexamethylenediamine (2,2,4- and 2,4,4- mixture)
EINECSNumber (2)-0154&(2)-3719
TSCA Yes