missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylene Sulfide 98.0+%, TCI America™
Supplier: TCI America T047425ML
Specifications
| Trimethylene Sulfide | |
| Yellow | |
| C3H6S | |
| 25 mL | |
| trimethylene sulfide, thiacyclobutane, propane, 1,3-epithio, unii-1o7c19qqdb, 1o7c19qqdb, 1,3-epithio propane, acmc-209h4c, trimethylene sulfide 8ci, trimethylene sulfide gc, inchi=1/c3h6s/c1-2-4-3-1/h1-3h | |
| C1CSC1 | |
| 74.14 | |
| CHEBI:87565 | |
| ≥98.0% (GC) |
| 287-27-4 | |
| 94°C | |
| MFCD00042751 | |
| 1993 | |
| XSROQCDVUIHRSI-UHFFFAOYSA-N | |
| thietane | |
| 9251 | |
| 74.14 | |
| Liquid |
Chemical Identifiers
| 287-27-4 | |
| 74.14 | |
| XSROQCDVUIHRSI-UHFFFAOYSA-N | |
| 9251 | |
| thietane |
| C3H6S | |
| MFCD00042751 | |
| trimethylene sulfide, thiacyclobutane, propane, 1,3-epithio, unii-1o7c19qqdb, 1o7c19qqdb, 1,3-epithio propane, acmc-209h4c, trimethylene sulfide 8ci, trimethylene sulfide gc, inchi=1/c3h6s/c1-2-4-3-1/h1-3h | |
| CHEBI:87565 | |
| C1CSC1 |
Safety and Handling
TSCA : Yes