missing translation for 'onlineSavingsMsg'
Learn More

Trimethylchlorogermane 98.0+%, TCI America™

Supplier:  TCI America T11611G

Catalog No. T11611G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Trimethylchlorogermane
Colorless
C3H9ClGe
1 g
trimethylchlorogermane, trimethylgermanium chloride, chloro trimethyl germane, chlorotrimethylgermanium, trimethylgermyl chloride, ch3 3gecl, acmc-209d8g, chlorotrimethylgermane
C[Ge](C)(C)Cl
153.185
153.19
Liquid
1529-47-1
102°C
MFCD00000462
2924
ZZBNZZCHSNOXOH-UHFFFAOYSA-N
chloro(trimethyl)germane
73723
≥98.0% (GC)

Chemical Identifiers

1529-47-1
153.185
ZZBNZZCHSNOXOH-UHFFFAOYSA-N
73723
C[Ge](C)(C)Cl
C3H9ClGe
MFCD00000462
trimethylchlorogermane, trimethylgermanium chloride, chloro trimethyl germane, chlorotrimethylgermanium, trimethylgermyl chloride, ch3 3gecl, acmc-209d8g, chlorotrimethylgermane
chloro(trimethyl)germane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions