missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine N-Oxide Anhydrous 95.0+%, TCI America™

Catalog No. p-7136735
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13621G 1 g
T13625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13621G Supplier TCI America Supplier No. T13621G
Only null left

Chemical Identifiers

CAS 1184-78-7
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.111
MDL Number MFCD00002048
InChI Key UYPYRKYUKCHHIB-UHFFFAOYSA-N
Synonym TMANO
PubChem CID 1145
ChEBI CHEBI:15724
IUPAC Name N,N-dimethylmethanamine oxide
SMILES C[N+](C)(C)[O-]

Specifications

Melting Point 222°C
Color White-Yellow
Quantity 1 g
Formula Weight 75.11
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylamine N-Oxide Anhydrous