missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7136296
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T046525G 25 g
T0465100G 100 g
T0465500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T046525G Supplier TCI America Supplier No. T046525G
Only null left

Chemical Identifiers

CAS 593-81-7
Molecular Formula C3H10ClN
Molecular Weight (g/mol) 95.57
MDL Number MFCD00012478
InChI Key SZYJELPVAFJOGJ-UHFFFAOYSA-N
Synonym trimethylamine hydrochloride, trimethylammonium chloride, trimethylamine hcl, n,n-dimethylmethanamine hydrochloride, trimethylamine monohydrochloride, methanamine, n,n-dimethyl-, hydrochloride, trimethylaminehydrochloride, unii-uw38srm77k, uw38srm77k, trimethyl ammonium chloride
PubChem CID 10313079
ChEBI CHEBI:64700
IUPAC Name hydrogen trimethylamine chloride
SMILES [H+].[Cl-].CN(C)C

Specifications

Color White
Quantity 25 g
Formula Weight 95.57
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylamine Hydrochloride
RTECSNumber YH2700000
TSCA Yes