missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine (ca. 13% in Tetrahydrofuran, ca. 2mol/L), TCI America™

Catalog No. p-7137365
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2704100ML 100 mL
T2704500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2704100ML Supplier TCI America Supplier No. T2704100ML
Only null left

Chemical Identifiers

CAS 75-50-3
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00008327
InChI Key GETQZCLCWQTVFV-UHFFFAOYSA-N
Synonym trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
PubChem CID 1146
ChEBI CHEBI:18139
IUPAC Name trimethylamine
SMILES CN(C)C

Specifications

Color Colorless
Quantity 100 mL
UN Number 2924
Formula Weight 59.11
Physical Form Liquid
Chemical Name or Material Trimethylamine (ca. 13% in Tetrahydrofuran, ca. 2mol/L)
EINECSNumber (2)-0140
RTECSNumber PA0350000
TSCA Yes
Recommended Storage Refrigerator