missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine Borane 97.0+%, TCI America™

Catalog No. T118125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T118125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T118125G Supplier TCI America Supplier No. T118125G

Chemical Identifiers

CAS 75-22-9
Molecular Formula C3H12BN
Molecular Weight (g/mol) 72.95
MDL Number MFCD00012420
InChI Key WVMHLYQJPRXKLC-UHFFFAOYSA-N
Synonym Borane - Trimethylamine Complex, TMAB
IUPAC Name trimethylamine borane
SMILES B.CN(C)C

Specifications

Melting Point 95°C
Color White
Quantity 25 g
UN Number 1325
Formula Weight 72.95
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylamine Borane
RTECSNumber PA0525000
TSCA Yes