missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Trimethylamine

Catalog No. 243205400G
Encompass
Change view
Click to view available options
Quantity:
400 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
243205400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 243205400G Supplier Sigma Aldrich Supplier No. 243205400G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trimethylamine

Specifications

CAS 75-50-3
Melting Point -117°C
Density 0.63 g/mL at 20°C
Boiling Point 3°C to 4°C
Molecular Formula C3H9N
Linear Formula (CH3)3N
MDL Number MFCD00008327
Quantity 400 g
Molecular Weight (g/mol) 59.11 g/mol