missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine, 1M soln. in THF

Catalog No. p-7031725
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32899AE 100 mL
AAH32899AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32899AE Supplier Thermo Scientific Chemicals Supplier No. H32899AE
Only null left

CAS: 75-50-3 | C3H9N | 59.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylamine complexes of alane as precursors for the low-pressure chemical vapor deposition of aluminum. Trimethylamine was found to be unsuitable as an agent for differentiating between Lewis and Brpinsted sites because it is dissociatively adsorbed and is, therefore useful for the perturbing effects. Choline derivative manufactured from TMA are used in medication of liver disorders. Used directly as acid binder in the manufacture of Sulpha Drugs. Animal/poultry feed additive like choline chloride is manufactured from TMA. Used in the manufacture of alkyl quaternary ammonium salts. To manufacture Ion exchange Resins based on chloromethylated polystyrene.

Solubility
Easily soluble in water, also soluble in most organic solvents (such as methanol and ethanol).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Chemical Identifiers

CAS 75-50-3
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00008327
InChI Key GETQZCLCWQTVFV-UHFFFAOYSA-N
Synonym trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241
PubChem CID 1146
ChEBI CHEBI:18139
IUPAC Name N,N-dimethylmethanamine
SMILES CN(C)C

Specifications

Density 0.85
Linear Formula (CH3)3N
Quantity 100 mL
UN Number UN1993
Sensitivity Hygroscopic
Solubility Information Easily soluble in water,also soluble in most organic solvents (such as methanol and ethanol).
Formula Weight 59.11
Concentration or Composition (by Analyte or Components) 1M soln. in THF
Chemical Name or Material Trimethylamine
Hazard Category H225-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H224-H318-H302-H332-H335-H315
Extremely flammable liquid and vapor.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 200-875-0
RTECSNumber PA0350000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only