Learn More
Trimethylacetylacetonitrile, 99%, Thermo Scientific Chemicals
Description
Trimethylacetylacetonitrile is a useful intermediate. 4,4-Dimethyl-3-oxopentanenitrile has been used in the preparation of 4,4-dimethyl-3-oxo-2-benzylpentanenitrile, 4,4-dimethyl-3-oxo-2-(4-hydroxybenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(4-methoxybenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(4-fluorobenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(3,4-methylenedioxybenzyl)pentanenitrile, 1-(5-tert-butyl-2-p-tolyl-2H-pyrazol-3-yl)-3-phenylurea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trimethylacetylacetonitrile is a useful intermediate. 4,4-Dimethyl-3-oxopentanenitrile has been used in the preparation of 4,4-dimethyl-3-oxo-2-benzylpentanenitrile, 4,4-dimethyl-3-oxo-2-(4-hydroxybenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(4-methoxybenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(4-fluorobenzyl)pentanenitrile, 4,4-dimethyl-3-oxo-2-(3,4-methylenedioxybenzyl)pentanenitrile, 1-(5-tert-butyl-2-p-tolyl-2H-pyrazol-3-yl)-3-phenylurea.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Specifications
Specifications
| Melting Point | 66°C to 70°C |
| Quantity | 25 g |
| UN Number | UN3439 |
| Beilstein | 1746674 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 125.17 |
| Percent Purity | 99% |
| Chemical Name or Material | Trimethylacetylacetonitrile |
Safety and Handling
| Hazard Category | H301-H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301 Toxic if swallowed. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S. |
| EINECSNumber | 262-017-1 |
| RTECSNumber | SA2991000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only