Learn More
Trimethylacetyl chloride, 98+%
Description
Trimethylacetyl chloride is used as a precursor in the preparation of tert-butyl peroxypivalate, guttiferon A derivatives, which is potential for the treatment of malaria. It is used as a raw material in the production of synthetic acidamide medicament and phenol ester medicament. In addition to this, it is used for the synthesis of active pharmaceutical ingredients such as aminobenzylpenicilin, cephalexin, cefazolin, dipivefrin and dipivalyl epinephrine. It is also used in heavy polymerization, N-acylating agent for amines, Schiff bases, pyrrolidinones as well as an O-acylating agent for alcohols, lactones and saccharides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Trimethylacetyl chloride is used as a precursor in the preparation of tert-butyl peroxypivalate, guttiferon A derivatives, which is potential for the treatment of malaria. It is used as a raw material in the production of synthetic acidamide medicament and phenol ester medicament. In addition to this, it is used for the synthesis of active pharmaceutical ingredients such as aminobenzylpenicilin, cephalexin, cefazolin, dipivefrin and dipivalyl epinephrine. It is also used in heavy polymerization, N-acylating agent for amines, Schiff bases, pyrrolidinones as well as an O-acylating agent for alcohols, lactones and saccharides.
Solubility
Miscible with acetonitrile.
Notes
Moisture sensitive. Incompatible withalcohols, oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | -57°C |
| Density | 0.985 |
| Boiling Point | 105°C to 106°C |
| Flash Point | 8°C (46°F) |
| Odor | Pungent |
| Refractive Index | 1.412 |
| Quantity | 100 mL |
| UN Number | UN2438 |
| Beilstein | 385668 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H225-H290-H302-H314-H330 |
| Hazard Statement | GHS H Statement H225-H330-H314-H318-H302 Highly flammable liquid and vapour. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TRIMETHYLACETYL CHLORIDE |
| EINECSNumber | 221-921-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only